Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606231 MIM: 615748 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APPL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APPL2 - adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251904.1 | 3298 | Silent Mutation | AAC,AAT | N,N 661 | NP_001238833.1 | |
NM_001251905.1 | 3298 | Silent Mutation | AAC,AAT | N,N 612 | NP_001238834.1 | |
NM_018171.3 | 3298 | Silent Mutation | AAC,AAT | N,N 655 | NP_060641.2 | |
XM_006719472.1 | 3298 | Silent Mutation | AAC,AAT | N,N 614 | XP_006719535.1 | |
XM_011538530.2 | 3298 | Silent Mutation | AAC,AAT | N,N 648 | XP_011536832.1 | |
XM_011538531.2 | 3298 | Silent Mutation | AAC,AAT | N,N 618 | XP_011536833.1 | |
XM_011538532.2 | 3298 | Silent Mutation | AAC,AAT | N,N 618 | XP_011536834.1 | |
XM_017019551.1 | 3298 | Silent Mutation | AAC,AAT | N,N 642 | XP_016875040.1 | |
XM_017019552.1 | 3298 | Silent Mutation | AAC,AAT | N,N 612 | XP_016875041.1 | |
XM_017019553.1 | 3298 | Silent Mutation | AAC,AAT | N,N 612 | XP_016875042.1 | |
XM_017019554.1 | 3298 | Silent Mutation | AAC,AAT | N,N 608 | XP_016875043.1 |
KIAA1033 - KIAA1033 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |