Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602008 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IPO5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IPO5 - importin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002271.4 | Intron | NP_002262.3 | ||||
XM_005254052.3 | Intron | XP_005254109.1 | ||||
XM_005254053.3 | Intron | XP_005254110.1 | ||||
XM_011521087.2 | Intron | XP_011519389.1 | ||||
XM_011521088.2 | Intron | XP_011519390.1 | ||||
XM_011521089.2 | Intron | XP_011519391.1 | ||||
XM_011521090.2 | Intron | XP_011519392.1 | ||||
XM_017020562.1 | Intron | XP_016876051.1 |
Set Membership: |
HapMap |