Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608275 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAB21L3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
MAB21L3 - mab-21 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152367.2 | Intron | NP_689580.2 | ||||
XM_011540623.2 | Intron | XP_011538925.1 | ||||
XM_011540625.2 | Intron | XP_011538927.1 | ||||
XM_011540626.2 | Intron | XP_011538928.1 | ||||
XM_017000260.1 | Intron | XP_016855749.1 |
SLC22A15 - solute carrier family 22 member 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |