Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601863 | ||||||||||||||||||||
Literature Links: |
GBAT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GBAT2 - glioblastoma multiforme-associated transcript 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RFX5 - regulatory factor X5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000449.3 | 2244 | UTR 3 | NP_000440.1 | |||
NM_001025603.1 | 2244 | UTR 3 | NP_001020774.1 | |||
XM_005245405.1 | 2244 | UTR 3 | XP_005245462.1 | |||
XM_005245406.3 | 2244 | UTR 3 | XP_005245463.1 | |||
XM_011509847.1 | 2244 | UTR 3 | XP_011508149.1 | |||
XM_011509848.1 | 2244 | UTR 3 | XP_011508150.1 | |||
XM_011509849.1 | 2244 | UTR 3 | XP_011508151.1 | |||
XM_011509850.1 | 2244 | UTR 3 | XP_011508152.1 | |||
XM_017001999.1 | 2244 | UTR 3 | XP_016857488.1 | |||
XM_017002000.1 | 2244 | UTR 3 | XP_016857489.1 |