Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
46 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612875 MIM: 603867 MIM: 605313 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNRHR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GNRHR2 - gonadotropin releasing hormone receptor 2 (pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEX11B - peroxisomal biogenesis factor 11 beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM8A - RNA binding motif protein 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005105.4 | Intron | NP_005096.1 |