Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605643 MIM: 602652 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
KLK5 - kallikrein related peptidase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLK6 - kallikrein related peptidase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012964.2 | 927 | Silent Mutation | CTC,CTG | L,L 162 | NP_001012982.1 | |
NM_001012965.2 | 927 | Silent Mutation | CTC,CTG | L,L 55 | NP_001012983.1 | |
NM_001319948.1 | 927 | Silent Mutation | CTC,CTG | L,L 55 | NP_001306877.1 | |
NM_001319949.1 | 927 | Silent Mutation | CTC,CTG | L,L 55 | NP_001306878.1 | |
NM_002774.3 | 927 | Silent Mutation | CTC,CTG | L,L 162 | NP_002765.1 |
Set Membership: |
HapMap Validated |