Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAGAAGAGCTGACAGGTCTGAA[G/T]GGGGGAGACTCAGTTATCCGGAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607419 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GEMIN7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GEMIN7 - gem nuclear organelle associated protein 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372419 - uncharacterized LOC105372419 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R37 - protein phosphatase 1 regulatory subunit 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019121.1 | Intron | NP_061994.1 | ||||
XM_006723159.2 | Intron | XP_006723222.1 |