Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606492 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EXOSC5 - exosome component 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM91 - transmembrane protein 91 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042595.2 | 275 | Intron | NP_001036060.1 | |||
NM_001098821.1 | 275 | UTR 5 | NP_001092291.1 | |||
NM_001098822.1 | 275 | UTR 5 | NP_001092292.1 | |||
NM_001098823.1 | 275 | UTR 5 | NP_001092293.1 | |||
NM_001098824.1 | 275 | UTR 5 | NP_001092294.1 | |||
NM_001098825.1 | 275 | Intron | NP_001092295.1 |