Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164176 | ||||||||||||||||||||
Literature Links: |
DEDD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DEDD2 - death effector domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270614.1 | 1667 | UTR 3 | NP_001257543.1 | |||
NM_001270615.1 | 1667 | UTR 3 | NP_001257544.1 | |||
NM_133328.3 | 1667 | UTR 3 | NP_579874.1 | |||
XM_011526569.1 | 1667 | UTR 3 | XP_011524871.1 | |||
XM_011526571.1 | 1667 | UTR 3 | XP_011524873.1 | |||
XM_011526572.1 | 1667 | UTR 3 | XP_011524874.1 | |||
XM_017026402.1 | 1667 | UTR 3 | XP_016881891.1 | |||
XM_017026403.1 | 1667 | UTR 3 | XP_016881892.1 |