Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606784 MIM: 614387 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEDD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
DEDD2 - death effector domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270614.1 | 363 | Intron | NP_001257543.1 | |||
NM_001270615.1 | 363 | Intron | NP_001257544.1 | |||
NM_133328.3 | 363 | Intron | NP_579874.1 | |||
XM_011526569.1 | 363 | Intron | XP_011524871.1 | |||
XM_011526571.1 | 363 | Intron | XP_011524873.1 | |||
XM_011526572.1 | 363 | Intron | XP_011524874.1 | |||
XM_017026402.1 | 363 | Intron | XP_016881891.1 | |||
XM_017026403.1 | 363 | Intron | XP_016881892.1 |
GSK3A - glycogen synthase kinase 3 alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF526 - zinc finger protein 526 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001314033.1 | 363 | UTR 5 | NP_001300962.1 | |||
NM_133444.2 | 363 | Intron | NP_597701.1 |