Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 109770 | ||||||||||||||||||||
Literature Links: |
CEACAM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CEACAM1 - carcinoembryonic antigen related cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024912.2 | 1922 | UTR 3 | NP_001020083.1 | |||
NM_001184813.1 | 1922 | UTR 3 | NP_001171742.1 | |||
NM_001184815.1 | 1922 | UTR 3 | NP_001171744.1 | |||
NM_001184816.1 | 1922 | UTR 3 | NP_001171745.1 | |||
NM_001205344.1 | 1922 | UTR 3 | NP_001192273.1 | |||
NM_001712.4 | 1922 | UTR 3 | NP_001703.2 | |||
XM_011527206.1 | 1922 | Intron | XP_011525508.1 |
LIPE-AS1 - LIPE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |