Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607320 MIM: 180901 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASGRP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RASGRP4 - RAS guanyl releasing protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RYR1 - ryanodine receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000540.2 | Intron | NP_000531.2 | ||||
NM_001042723.1 | Intron | NP_001036188.1 | ||||
XM_006723317.2 | Intron | XP_006723380.1 | ||||
XM_006723319.2 | Intron | XP_006723382.1 | ||||
XM_011527205.2 | Intron | XP_011525507.1 |