Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614567 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIAPH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DIAPH3 - diaphanous related formin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042517.1 | Intron | NP_001035982.1 | ||||
NM_001258366.1 | Intron | NP_001245295.1 | ||||
NM_001258367.1 | Intron | NP_001245296.1 | ||||
NM_001258368.1 | Intron | NP_001245297.1 | ||||
NM_001258369.1 | Intron | NP_001245298.1 | ||||
NM_001258370.1 | Intron | NP_001245299.1 | ||||
NM_030932.3 | Intron | NP_112194.2 | ||||
XM_006719876.1 | Intron | XP_006719939.1 | ||||
XM_011535258.2 | Intron | XP_011533560.1 | ||||
XM_011535263.2 | Intron | XP_011533565.1 | ||||
XM_011535265.2 | Intron | XP_011533567.1 | ||||
XM_017020789.1 | Intron | XP_016876278.1 |