Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF23 - immunoglobulin superfamily member 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205280.1 | Intron | NP_001192209.1 | ||||
XM_017026317.1 | Intron | XP_016881806.1 | ||||
XM_017026318.1 | Intron | XP_016881807.1 | ||||
XM_017026319.1 | Intron | XP_016881808.1 | ||||
XM_017026320.1 | Intron | XP_016881809.1 | ||||
XM_017026321.1 | Intron | XP_016881810.1 | ||||
XM_017026322.1 | Intron | XP_016881811.1 | ||||
XM_017026323.1 | Intron | XP_016881812.1 | ||||
XM_017026324.1 | Intron | XP_016881813.1 | ||||
XM_017026325.1 | Intron | XP_016881814.1 | ||||
XM_017026326.1 | Intron | XP_016881815.1 |