Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613176 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMG9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SMG9 - SMG9, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005259057.2 | Intron | XP_005259114.1 | ||||
XM_011527113.1 | Intron | XP_011525415.1 | ||||
XM_011527114.1 | Intron | XP_011525416.1 | ||||
XM_011527115.1 | Intron | XP_011525417.1 | ||||
XM_011527116.1 | Intron | XP_011525418.1 | ||||
XM_017026988.1 | Intron | XP_016882477.1 |