Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602566 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RX7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
P2RX7 - purinergic receptor P2X 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002562.5 | Intron | NP_002553.3 | ||||
XM_011538419.2 | Intron | XP_011536721.1 | ||||
XM_011538420.2 | Intron | XP_011536722.1 | ||||
XM_017019364.1 | Intron | XP_016874853.1 | ||||
XM_017019365.1 | Intron | XP_016874854.1 | ||||
XM_017019366.1 | Intron | XP_016874855.1 | ||||
XM_017019367.1 | Intron | XP_016874856.1 |
Set Membership: |
HapMap |