Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 160775 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6819 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6819 - microRNA 6819 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYH9 - myosin, heavy chain 9, non-muscle | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002473.5 | 6650 | UTR 3 | NP_002464.1 | |||
XM_011530197.2 | 6650 | UTR 3 | XP_011528499.1 | |||
XM_017028803.1 | 6650 | UTR 3 | XP_016884292.1 | |||
XM_017028804.1 | 6650 | UTR 3 | XP_016884293.1 | |||
XM_017028805.1 | 6650 | UTR 3 | XP_016884294.1 | |||
XM_017028806.1 | 6650 | UTR 3 | XP_016884295.1 |