Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107325 MIM: 126380 MIM: 607463 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD3EAP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD3EAP - CD3e molecule associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297590.1 | 548 | UTR 5 | NP_001284519.1 | |||
NM_012099.1 | 548 | UTR 5 | NP_036231.1 | |||
XM_017026180.1 | 548 | UTR 5 | XP_016881669.1 | |||
XM_017026181.1 | 548 | UTR 5 | XP_016881670.1 |
ERCC1 - ERCC excision repair 1, endonuclease non-catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166049.1 | 548 | Intron | NP_001159521.1 | |||
NM_001983.3 | 548 | Intron | NP_001974.1 | |||
NM_202001.2 | 548 | Intron | NP_973730.1 | |||
XM_005258634.1 | 548 | Intron | XP_005258691.1 | |||
XM_005258635.2 | 548 | Intron | XP_005258692.1 | |||
XM_005258636.4 | 548 | Intron | XP_005258693.1 | |||
XM_005258637.1 | 548 | Intron | XP_005258694.1 | |||
XM_011526610.2 | 548 | Intron | XP_011524912.1 | |||
XM_017026459.1 | 548 | Intron | XP_016881948.1 | |||
XM_017026460.1 | 548 | Intron | XP_016881949.1 | |||
XM_017026461.1 | 548 | Intron | XP_016881950.1 | |||
XM_017026462.1 | 548 | Intron | XP_016881951.1 | |||
XM_017026463.1 | 548 | Intron | XP_016881952.1 | |||
XM_017026464.1 | 548 | Intron | XP_016881953.1 | |||
XM_017026465.1 | 548 | Intron | XP_016881954.1 | |||
XM_017026466.1 | 548 | Intron | XP_016881955.1 |
PPP1R13L - protein phosphatase 1 regulatory subunit 13 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142502.1 | 548 | Intron | NP_001135974.1 | |||
NM_006663.3 | 548 | Intron | NP_006654.2 | |||
XM_017026177.1 | 548 | Intron | XP_016881666.1 | |||
XM_017026178.1 | 548 | Intron | XP_016881667.1 | |||
XM_017026179.1 | 548 | Intron | XP_016881668.1 |
Set Membership: |
Validated |