Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606882 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP7B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP7B - ATPase copper transporting beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000053.3 | 5716 | UTR 3 | NP_000044.2 | |||
NM_001005918.2 | 5716 | UTR 3 | NP_001005918.1 | |||
NM_001243182.1 | 5716 | UTR 3 | NP_001230111.1 | |||
XM_005266423.2 | 5716 | UTR 3 | XP_005266480.1 | |||
XM_005266424.4 | 5716 | UTR 3 | XP_005266481.1 | |||
XM_005266427.2 | 5716 | UTR 3 | XP_005266484.1 | |||
XM_005266428.1 | 5716 | UTR 3 | XP_005266485.1 | |||
XM_005266430.4 | 5716 | UTR 3 | XP_005266487.1 | |||
XM_005266431.3 | 5716 | UTR 3 | XP_005266488.1 | |||
XM_005266432.2 | 5716 | UTR 3 | XP_005266489.1 | |||
XM_006719837.3 | 5716 | UTR 3 | XP_006719900.1 | |||
XM_006719838.1 | 5716 | UTR 3 | XP_006719901.1 | |||
XM_006719839.1 | 5716 | UTR 3 | XP_006719902.1 | |||
XM_011535117.2 | 5716 | UTR 3 | XP_011533419.1 | |||
XM_011535118.1 | 5716 | UTR 3 | XP_011533420.1 | |||
XM_011535119.1 | 5716 | UTR 3 | XP_011533421.1 | |||
XM_011535122.2 | 5716 | UTR 3 | XP_011533424.1 | |||
XM_017020627.1 | 5716 | UTR 3 | XP_016876116.1 | |||
XM_017020628.1 | 5716 | UTR 3 | XP_016876117.1 |
Set Membership: |
DME Validated Inventoried |