Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131560 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLOT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FLOT2 - flotillin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004475.2 | 1470 | UTR 3 | NP_004466.2 | |||
XM_005257950.3 | 1470 | UTR 3 | XP_005258007.1 | |||
XM_005257953.2 | 1470 | UTR 3 | XP_005258010.1 | |||
XM_017024394.1 | 1470 | UTR 3 | XP_016879883.1 | |||
XM_017024395.1 | 1470 | UTR 3 | XP_016879884.1 | |||
XM_017024396.1 | 1470 | UTR 3 | XP_016879885.1 |