Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602464 | ||||||||||||||||||||
Literature Links: |
FAM222B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM222B - family with sequence similarity 222 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077498.2 | 4261 | UTR 3 | NP_001070966.1 | |||
NM_001288631.1 | 4261 | UTR 3 | NP_001275560.1 | |||
NM_001288632.1 | 4261 | UTR 3 | NP_001275561.1 | |||
NM_001288633.1 | 4261 | UTR 3 | NP_001275562.1 | |||
NM_001288634.1 | 4261 | UTR 3 | NP_001275563.1 | |||
NM_001288635.1 | 4261 | UTR 3 | NP_001275564.1 | |||
NM_001288636.1 | 4261 | UTR 3 | NP_001275565.1 | |||
NM_001288637.1 | 4261 | UTR 3 | NP_001275566.1 | |||
NM_001288638.1 | 4261 | UTR 3 | NP_001275567.1 | |||
NM_001288639.1 | 4261 | UTR 3 | NP_001275568.1 | |||
NM_001288640.1 | 4261 | UTR 3 | NP_001275569.1 | |||
NM_018182.3 | 4261 | UTR 3 | NP_060652.2 | |||
XM_011524986.2 | 4261 | UTR 3 | XP_011523288.1 | |||
XM_011524987.1 | 4261 | UTR 3 | XP_011523289.1 | |||
XM_011524988.2 | 4261 | UTR 3 | XP_011523290.1 | |||
XM_017024831.1 | 4261 | UTR 3 | XP_016880320.1 | |||
XM_017024832.1 | 4261 | UTR 3 | XP_016880321.1 | |||
XM_017024833.1 | 4261 | UTR 3 | XP_016880322.1 | |||
XM_017024834.1 | 4261 | UTR 3 | XP_016880323.1 | |||
XM_017024835.1 | 4261 | UTR 3 | XP_016880324.1 |
TRAF4 - TNF receptor associated factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |