Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615941 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HACD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HACD4 - 3-hydroxyacyl-CoA dehydratase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010915.4 | 1229 | UTR 3 | NP_001010915.2 | |||
NM_001321883.1 | 1229 | UTR 3 | NP_001308812.1 | |||
NM_001321903.1 | 1229 | UTR 3 | NP_001308832.1 | |||
XM_011517876.2 | 1229 | UTR 3 | XP_011516178.1 | |||
XM_017014713.1 | 1229 | UTR 3 | XP_016870202.1 | |||
XM_017014714.1 | 1229 | UTR 3 | XP_016870203.1 |