Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGCCTGAGGACCTGGACGGCAT[A/T]GGCATGGGCCGGCCTGGTGAGGGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607611 MIM: 608076 | ||||||||||||||||||||
Literature Links: |
PLSCR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLSCR3 - phospholipid scramblase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM256-PLSCR3 - TMEM256-PLSCR3 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNK1 - tyrosine kinase non receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251902.1 | 379 | Silent Mutation | ATA,ATT | I,I 49 | NP_001238831.1 | |
NM_003985.4 | 379 | Silent Mutation | ATA,ATT | I,I 49 | NP_003976.2 | |
XM_011524045.2 | 379 | Silent Mutation | ATA,ATT | I,I 49 | XP_011522347.1 | |
XM_011524046.2 | 379 | Silent Mutation | ATA,ATT | I,I 49 | XP_011522348.1 | |
XM_011524047.2 | 379 | Silent Mutation | ATA,ATT | I,I 49 | XP_011522349.1 | |
XM_011524048.1 | 379 | UTR 5 | XP_011522350.1 |