Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602293 MIM: 617029 | ||||||||||||||||||||
Literature Links: |
CIB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CIB1 - calcium and integrin binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277764.1 | 757 | UTR 3 | NP_001264693.1 | |||
NM_006384.3 | 757 | UTR 3 | NP_006375.2 | |||
XM_006720375.1 | 757 | Intron | XP_006720438.1 |
GDPGP1 - GDP-D-glucose phosphorylase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA4B - semaphorin 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324029.1 | 757 | Intron | NP_001310958.1 | |||
NM_001324030.1 | 757 | Intron | NP_001310959.1 | |||
NM_001324031.1 | 757 | Intron | NP_001310960.1 | |||
NM_001324032.1 | 757 | Intron | NP_001310961.1 | |||
NM_001324033.1 | 757 | Intron | NP_001310962.1 | |||
NM_001324034.1 | 757 | Intron | NP_001310963.1 | |||
NM_020210.3 | 757 | Intron | NP_064595.2 | |||
NM_198925.2 | 757 | Intron | NP_945119.1 |