Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611411 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMKK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAMKK1 - calcium/calmodulin dependent protein kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032294.2 | Intron | NP_115670.1 | ||||
NM_172206.1 | Intron | NP_757343.1 | ||||
NM_172207.2 | Intron | NP_757344.2 | ||||
XM_005256817.1 | Intron | XP_005256874.1 | ||||
XM_006721588.2 | Intron | XP_006721651.1 | ||||
XM_011524031.2 | Intron | XP_011522333.2 | ||||
XM_011524032.2 | Intron | XP_011522334.1 | ||||
XM_011524033.1 | Intron | XP_011522335.1 | ||||
XM_011524035.2 | Intron | XP_011522337.1 | ||||
XM_017025219.1 | Intron | XP_016880708.1 | ||||
XM_017025220.1 | Intron | XP_016880709.1 | ||||
XM_017025221.1 | Intron | XP_016880710.1 | ||||
XM_017025222.1 | Intron | XP_016880711.1 |