Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602552 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP88 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUP88 - nucleoporin 88 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320653.1 | 309 | Intron | NP_001307582.1 | |||
NM_002532.5 | 309 | Intron | NP_002523.2 | |||
XM_011523893.2 | 309 | Intron | XP_011522195.1 | |||
XM_017024694.1 | 309 | Intron | XP_016880183.1 |
RPAIN - RPA interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033002.3 | 309 | UTR 5 | NP_001028174.2 | |||
NM_001160243.1 | 309 | UTR 5 | NP_001153715.1 | |||
NM_001160244.1 | 309 | UTR 5 | NP_001153716.1 | |||
NM_001160246.1 | 309 | UTR 5 | NP_001153718.1 | |||
NM_001160266.1 | 309 | UTR 5 | NP_001153738.1 |
Set Membership: |
HapMap |