Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGATATTTGTTTCTCCAGTTTT[C/T]TGGCTCTAAGAAATTACACTTTCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610304 MIM: 614405 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DERL2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DERL2 - derlin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304777.1 | 1014 | UTR 3 | NP_001291706.1 | |||
NM_001304779.1 | 1014 | UTR 3 | NP_001291708.1 | |||
NM_016041.4 | 1014 | UTR 3 | NP_057125.2 |
DHX33 - DEAH-box helicase 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105371506 - uncharacterized LOC105371506 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |