Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614787 MIM: 602177 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POGZ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POGZ - pogo transposable element with ZNF domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001194937.1 | 4978 | Silent Mutation | NP_001181866.1 | |||
NM_001194938.1 | 4978 | Silent Mutation | NP_001181867.1 | |||
NM_015100.3 | 4978 | Silent Mutation | NP_055915.2 | |||
NM_145796.3 | 4978 | Silent Mutation | NP_665739.3 | |||
NM_207171.2 | 4978 | Silent Mutation | NP_997054.1 | |||
XM_005244999.2 | 4978 | Silent Mutation | XP_005245056.1 | |||
XM_005245000.4 | 4978 | Silent Mutation | XP_005245057.1 | |||
XM_005245001.2 | 4978 | Silent Mutation | XP_005245058.1 | |||
XM_005245005.1 | 4978 | Silent Mutation | XP_005245062.1 | |||
XM_005245006.4 | 4978 | Silent Mutation | XP_005245063.1 | |||
XM_011509331.2 | 4978 | Silent Mutation | XP_011507633.1 | |||
XM_017000744.1 | 4978 | Silent Mutation | XP_016856233.1 | |||
XM_017000745.1 | 4978 | Silent Mutation | XP_016856234.1 | |||
XM_017000746.1 | 4978 | Silent Mutation | XP_016856235.1 | |||
XM_017000747.1 | 4978 | Silent Mutation | XP_016856236.1 | |||
XM_017000748.1 | 4978 | Silent Mutation | XP_016856237.1 | |||
XM_017000749.1 | 4978 | Silent Mutation | XP_016856238.1 |
PSMB4 - proteasome subunit beta 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |