Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603866 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PEX11A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PEX11A - peroxisomal biogenesis factor 11 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271572.1 | 11 | UTR 5 | NP_001258501.1 | |||
NM_001271573.1 | 11 | UTR 5 | NP_001258502.1 | |||
NM_003847.2 | 11 | UTR 5 | NP_003838.1 |
WDR93 - WD repeat domain 93 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284395.1 | 11 | Intron | NP_001271324.1 | |||
NM_001284396.1 | 11 | Intron | NP_001271325.1 | |||
NM_020212.1 | 11 | Intron | NP_064597.1 | |||
XM_006720614.1 | 11 | Intron | XP_006720677.1 | |||
XM_006720615.1 | 11 | Intron | XP_006720678.1 | |||
XM_006720618.2 | 11 | Intron | XP_006720681.1 | |||
XM_011521794.2 | 11 | Intron | XP_011520096.1 | |||
XM_011521795.1 | 11 | Intron | XP_011520097.1 | |||
XM_011521796.2 | 11 | Intron | XP_011520098.1 | |||
XM_011521798.2 | 11 | Intron | XP_011520100.1 | |||
XM_011521800.1 | 11 | Intron | XP_011520102.1 | |||
XM_011521802.2 | 11 | Intron | XP_011520104.1 | |||
XM_017022415.1 | 11 | Intron | XP_016877904.1 | |||
XM_017022416.1 | 11 | Intron | XP_016877905.1 | |||
XM_017022417.1 | 11 | Intron | XP_016877906.1 |
Set Membership: |
HapMap |