Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611479 MIM: 612762 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GPN1 - GPN-loop GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145047.1 | 1536 | UTR 3 | NP_001138519.1 | |||
NM_001145048.1 | 1536 | UTR 3 | NP_001138520.1 | |||
NM_001145049.1 | 1536 | UTR 3 | NP_001138521.1 | |||
NM_007266.3 | 1536 | UTR 3 | NP_009197.2 |
SUPT7L - SPT7-like STAGA complex gamma subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282729.1 | 1536 | Intron | NP_001269658.1 | |||
NM_001282730.1 | 1536 | Intron | NP_001269659.1 | |||
NM_001282731.1 | 1536 | Intron | NP_001269660.1 | |||
NM_001282732.1 | 1536 | Intron | NP_001269661.1 | |||
NM_014860.2 | 1536 | Intron | NP_055675.1 | |||
XM_005264672.4 | 1536 | Intron | XP_005264729.1 |
Set Membership: |
HapMap Validated |