Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610646 | ||||||||||||||||||||
Literature Links: |
C2orf70 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf70 - chromosome 2 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105519.2 | 998 | Intron | NP_001098989.1 | |||
NM_001322426.1 | 998 | Intron | NP_001309355.1 | |||
XM_011532810.2 | 998 | Intron | XP_011531112.1 | |||
XM_011532811.2 | 998 | UTR 5 | XP_011531113.1 | |||
XM_017003962.1 | 998 | Intron | XP_016859451.1 | |||
XM_017003963.1 | 998 | Intron | XP_016859452.1 | |||
XM_017003964.1 | 998 | Intron | XP_016859453.1 | |||
XM_017003965.1 | 998 | Intron | XP_016859454.1 | |||
XM_017003966.1 | 998 | Intron | XP_016859455.1 |
CIB4 - calcium and integrin binding family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |