Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCTTGAAACCAAGGTAACCAGC[A/G]TCAATATAATGAATCCCACAGAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604883 MIM: 137960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF3C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GTF3C2 - general transcription factor IIIC subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035521.2 | 2813 | Silent Mutation | GAC,GAT | D,D 690 | NP_001030598.1 | |
NM_001318909.1 | 2813 | Silent Mutation | GAC,GAT | D,D 701 | NP_001305838.1 | |
NM_001521.3 | 2813 | Silent Mutation | GAC,GAT | D,D 690 | NP_001512.1 | |
XM_005264272.4 | 2813 | Silent Mutation | GAC,GAT | D,D 729 | XP_005264329.1 | |
XM_011532801.2 | 2813 | Silent Mutation | GAC,GAT | D,D 805 | XP_011531103.1 |
GTF3C2-AS1 - GTF3C2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MPV17 - MPV17, mitochondrial inner membrane protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |