Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MFSD2B - major facilitator superfamily domain containing 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080473.1 | Intron | NP_001073942.1 | ||||
XM_005264301.4 | Intron | XP_005264358.1 | ||||
XM_017004063.1 | Intron | XP_016859552.1 | ||||
XM_017004064.1 | Intron | XP_016859553.1 | ||||
XM_017004065.1 | Intron | XP_016859554.1 | ||||
XM_017004066.1 | Intron | XP_016859555.1 | ||||
XM_017004067.1 | Intron | XP_016859556.1 | ||||
XM_017004068.1 | Intron | XP_016859557.1 | ||||
XM_017004069.1 | Intron | XP_016859558.1 | ||||
XM_017004070.1 | Intron | XP_016859559.1 |