Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615276 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CERS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CERS3 - ceramide synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290341.2 | 1660 | UTR 3 | NP_001277270.1 | |||
NM_001290342.2 | 1660 | UTR 3 | NP_001277271.1 | |||
NM_001290343.1 | 1660 | UTR 3 | NP_001277272.1 | |||
NM_178842.4 | 1660 | UTR 3 | NP_849164.2 | |||
XM_011521355.2 | 1660 | UTR 3 | XP_011519657.1 | |||
XM_011521357.2 | 1660 | UTR 3 | XP_011519659.1 | |||
XM_011521358.1 | 1660 | UTR 3 | XP_011519660.1 | |||
XM_017022002.1 | 1660 | UTR 3 | XP_016877491.1 | |||
XM_017022003.1 | 1660 | UTR 3 | XP_016877492.1 | |||
XM_017022004.1 | 1660 | UTR 3 | XP_016877493.1 |
CERS3-AS1 - CERS3 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |