Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607386 MIM: 606010 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT172 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
IFT172 - intraflagellar transport 172 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTCAP3 - keratinocyte associated protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRBP1 - nuclear receptor binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321357.1 | Intron | NP_001308286.1 | ||||
NM_001321358.1 | Intron | NP_001308287.1 | ||||
NM_001321359.1 | Intron | NP_001308288.1 | ||||
NM_001321361.1 | Intron | NP_001308290.1 | ||||
NM_001321362.1 | Intron | NP_001308291.1 | ||||
NM_001321363.1 | Intron | NP_001308292.1 | ||||
NM_013392.3 | Intron | NP_037524.1 |
Set Membership: |
HapMap Validated |