Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612157 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SENP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SENP1 - SUMO1/sentrin specific peptidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267594.1 | Intron | NP_001254523.1 | ||||
NM_001267595.1 | Intron | NP_001254524.1 | ||||
XM_006719361.3 | Intron | XP_006719424.1 | ||||
XM_006719362.2 | Intron | XP_006719425.1 | ||||
XM_011538244.2 | Intron | XP_011536546.1 | ||||
XM_011538245.2 | Intron | XP_011536547.1 | ||||
XM_017019229.1 | Intron | XP_016874718.1 | ||||
XM_017019230.1 | Intron | XP_016874719.1 | ||||
XM_017019231.1 | Intron | XP_016874720.1 | ||||
XM_017019232.1 | Intron | XP_016874721.1 | ||||
XM_017019233.1 | Intron | XP_016874722.1 | ||||
XM_017019234.1 | Intron | XP_016874723.1 | ||||
XM_017019235.1 | Intron | XP_016874724.1 | ||||
XM_017019236.1 | Intron | XP_016874725.1 | ||||
XM_017019237.1 | Intron | XP_016874726.1 | ||||
XM_017019238.1 | Intron | XP_016874727.1 | ||||
XM_017019239.1 | Intron | XP_016874728.1 | ||||
XM_017019240.1 | Intron | XP_016874729.1 |
Set Membership: |
HapMap |