Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605557 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00982 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LINC00982 - long intergenic non-protein coding RNA 982 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRDM16 - PR domain 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022114.3 | Intron | NP_071397.3 | ||||
NM_199454.2 | Intron | NP_955533.2 | ||||
XM_005244772.4 | Intron | XP_005244829.1 | ||||
XM_005244773.4 | Intron | XP_005244830.1 | ||||
XM_005244774.4 | Intron | XP_005244831.1 | ||||
XM_006710814.3 | Intron | XP_006710877.1 | ||||
XM_011541945.2 | Intron | XP_011540247.1 | ||||
XM_017002050.1 | Intron | XP_016857539.1 |
Set Membership: |
Validated |