Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 152310 | ||||||||||||||||||||
Literature Links: |
TFPI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TFPI - tissue factor pathway inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246818.1 | 2509 | UTR 3 | XP_005246875.1 | |||
XM_005246819.1 | 2509 | UTR 3 | XP_005246876.1 | |||
XM_006712720.3 | 2509 | UTR 3 | XP_006712783.1 | |||
XM_011511707.2 | 2509 | UTR 3 | XP_011510009.1 | |||
XM_011511708.1 | 2509 | UTR 3 | XP_011510010.1 | |||
XM_011511709.2 | 2509 | UTR 3 | XP_011510011.1 |