Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609394 | ||||||||||||||||||||
Literature Links: |
SPC24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPC24 - SPC24, NDC80 kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317031.1 | 1829 | UTR 3 | NP_001303960.1 | |||
NM_001317032.1 | 1829 | Intron | NP_001303961.1 | |||
NM_001317033.1 | 1829 | UTR 3 | NP_001303962.1 | |||
NM_182513.3 | 1829 | UTR 3 | NP_872319.1 | |||
XM_005259753.3 | 1829 | Intron | XP_005259810.1 | |||
XM_011527702.1 | 1829 | UTR 3 | XP_011526004.1 |