Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611463 MIM: 182205 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SAT2 - spermidine/spermine N1-acetyltransferase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320845.1 | Intron | NP_001307774.1 | ||||
NM_001320846.1 | Intron | NP_001307775.1 | ||||
NM_001320847.1 | Intron | NP_001307776.1 | ||||
NM_133491.4 | Intron | NP_597998.1 | ||||
XM_017024073.1 | Intron | XP_016879562.1 | ||||
XM_017024074.1 | Intron | XP_016879563.1 |
SHBG - sex hormone binding globulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040.4 | Intron | NP_001031.2 | ||||
NM_001146279.2 | Intron | NP_001139751.1 | ||||
NM_001146280.2 | Intron | NP_001139752.1 | ||||
NM_001146281.2 | Intron | NP_001139753.1 | ||||
NM_001289113.1 | Intron | NP_001276042.1 | ||||
NM_001289114.1 | Intron | NP_001276043.1 | ||||
NM_001289115.1 | Intron | NP_001276044.1 | ||||
NM_001289116.1 | Intron | NP_001276045.1 |
Set Membership: |
HapMap Validated |