Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 191170 | ||||||||||||||||||||
Literature Links: |
TP53 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TP53 - tumor protein p53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000546.5 | 1638 | UTR 3 | NP_000537.3 | |||
NM_001126112.2 | 1638 | UTR 3 | NP_001119584.1 | |||
NM_001126113.2 | 1638 | UTR 3 | NP_001119585.1 | |||
NM_001126114.2 | 1638 | UTR 3 | NP_001119586.1 | |||
NM_001126115.1 | 1638 | UTR 3 | NP_001119587.1 | |||
NM_001126116.1 | 1638 | UTR 3 | NP_001119588.1 | |||
NM_001126117.1 | 1638 | UTR 3 | NP_001119589.1 | |||
NM_001126118.1 | 1638 | UTR 3 | NP_001119590.1 | |||
NM_001276695.1 | 1638 | UTR 3 | NP_001263624.1 | |||
NM_001276696.1 | 1638 | UTR 3 | NP_001263625.1 | |||
NM_001276697.1 | 1638 | UTR 3 | NP_001263626.1 | |||
NM_001276698.1 | 1638 | UTR 3 | NP_001263627.1 | |||
NM_001276699.1 | 1638 | UTR 3 | NP_001263628.1 | |||
NM_001276760.1 | 1638 | UTR 3 | NP_001263689.1 | |||
NM_001276761.1 | 1638 | UTR 3 | NP_001263690.1 |