Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608801 MIM: 611487 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCDH PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GCDH - glutaryl-CoA dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000159.3 | 1713 | UTR 3 | NP_000150.1 | |||
NM_013976.3 | 1713 | UTR 3 | NP_039663.1 | |||
XM_006722721.2 | 1713 | Intron | XP_006722784.1 | |||
XM_011527899.2 | 1713 | Intron | XP_011526201.1 | |||
XM_011527900.2 | 1713 | Intron | XP_011526202.1 | |||
XM_017026580.1 | 1713 | Intron | XP_016882069.1 |
SYCE2 - synaptonemal complex central element protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105578.1 | 1713 | Intron | NP_001099048.1 | |||
XM_005259848.4 | 1713 | Intron | XP_005259905.1 | |||
XM_011527882.2 | 1713 | Intron | XP_011526184.1 | |||
XM_011527883.2 | 1713 | Intron | XP_011526185.1 |
Set Membership: |
HapMap Validated |