Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604708 MIM: 125860 | ||||||||||||||||||||||||||||||||
Literature Links: |
NFAT5 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
NFAT5 - nuclear factor of activated T-cells 5, tonicity-responsive | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NQO1 - NAD(P)H quinone dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000903.2 | 1019 | UTR 3 | NP_000894.1 | |||
NM_001025433.1 | 1019 | UTR 3 | NP_001020604.1 | |||
NM_001025434.1 | 1019 | UTR 3 | NP_001020605.1 | |||
NM_001286137.1 | 1019 | UTR 3 | NP_001273066.1 |
Set Membership: |
DME Validated Inventoried |