Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604708 MIM: 125860 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFAT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
NFAT5 - nuclear factor of activated T-cells 5, tonicity-responsive | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NQO1 - NAD(P)H quinone dehydrogenase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000903.2 | 2033 | UTR 3 | NP_000894.1 | |||
NM_001025433.1 | 2033 | UTR 3 | NP_001020604.1 | |||
NM_001025434.1 | 2033 | UTR 3 | NP_001020605.1 | |||
NM_001286137.1 | 2033 | UTR 3 | NP_001273066.1 |
Set Membership: |
HapMap DME Validated Inventoried |