Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603527 MIM: 610016 MIM: 613487 MIM: 607896 | ||||||||||||||||||||
Literature Links: |
DPH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DPH1 - diphthamide biosynthesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001383.3 | 1701 | UTR 3 | NP_001374.3 |
MIR132 - microRNA 132 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR212 - microRNA 212 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OVCA2 - ovarian tumor suppressor candidate 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080822.2 | 1701 | Missense Mutation | CAT,TAT | H,Y 173 | NP_543012.1 |