Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603825 MIM: 610963 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIC1 - hypermethylated in cancer 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMG6 - SMG6, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256827.1 | 4902 | UTR 3 | NP_001243756.1 | |||
NM_001256828.1 | 4902 | UTR 3 | NP_001243757.1 | |||
NM_001282326.1 | 4902 | UTR 3 | NP_001269255.1 | |||
NM_017575.4 | 4902 | UTR 3 | NP_060045.4 | |||
XM_005256569.3 | 4902 | UTR 3 | XP_005256626.1 | |||
XM_005256570.3 | 4902 | Intron | XP_005256627.1 | |||
XM_005256571.4 | 4902 | UTR 3 | XP_005256628.1 | |||
XM_011523769.2 | 4902 | UTR 3 | XP_011522071.1 | |||
XM_011523772.2 | 4902 | Intron | XP_011522074.1 | |||
XM_011523773.2 | 4902 | Intron | XP_011522075.1 | |||
XM_011523774.2 | 4902 | Intron | XP_011522076.1 | |||
XM_011523775.2 | 4902 | UTR 3 | XP_011522077.1 | |||
XM_017024398.1 | 4902 | Intron | XP_016879887.1 | |||
XM_017024399.1 | 4902 | UTR 3 | XP_016879888.1 |
Set Membership: |
HapMap |