Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300380 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTPS2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CTPS2 - CTP synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144002.1 | Intron | NP_001137474.1 | ||||
NM_019857.4 | Intron | NP_062831.3 | ||||
NM_175859.2 | Intron | NP_787055.1 | ||||
XM_005274562.3 | Intron | XP_005274619.1 | ||||
XM_006724503.3 | Intron | XP_006724566.1 | ||||
XM_011545545.2 | Intron | XP_011543847.1 |
Set Membership: |
HapMap |