Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118888 MIM: 606967 MIM: 177015 MIM: 176847 MIM: 604119 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTRL PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CTRL - chymotrypsin like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LCAT - lecithin-cholesterol acyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSKH1 - protein serine kinase H1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMB10 - proteasome subunit beta 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002801.3 | 82 | UTR 5 | NP_002792.1 |
SLC12A4 - solute carrier family 12 member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |