Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610458 MIM: 608700 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LZIC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LZIC - leucine zipper and CTNNBIP1 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316973.1 | Intron | NP_001303902.1 | ||||
NM_001316974.1 | Intron | NP_001303903.1 | ||||
NM_001316975.1 | Intron | NP_001303904.1 | ||||
NM_001316976.1 | Intron | NP_001303905.1 | ||||
NM_032368.4 | Intron | NP_115744.2 | ||||
XM_005263506.2 | Intron | XP_005263563.1 | ||||
XM_005263509.2 | Intron | XP_005263566.1 | ||||
XM_017002546.1 | Intron | XP_016858035.1 | ||||
XM_017002547.1 | Intron | XP_016858036.1 |
NMNAT1 - nicotinamide nucleotide adenylyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |