Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601826 MIM: 181031 | ||||||||||||||||||||
Literature Links: |
DGKD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DGKD - diacylglycerol kinase delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003648.2 | 294 | Intron | NP_003639.2 | |||
NM_152879.2 | 294 | Intron | NP_690618.2 | |||
XM_005246107.3 | 294 | Intron | XP_005246164.1 | |||
XM_005246108.1 | 294 | Intron | XP_005246165.1 | |||
XM_005246110.3 | 294 | Intron | XP_005246167.1 | |||
XM_011512028.2 | 294 | Intron | XP_011510330.1 | |||
XM_011512029.2 | 294 | Intron | XP_011510331.1 | |||
XM_011512030.2 | 294 | Intron | XP_011510332.1 | |||
XM_011512031.2 | 294 | Intron | XP_011510333.1 | |||
XM_011512032.2 | 294 | Intron | XP_011510334.1 | |||
XM_011512033.2 | 294 | Intron | XP_011510335.1 | |||
XM_011512035.1 | 294 | Intron | XP_011510337.1 | |||
XM_011512037.2 | 294 | Intron | XP_011510339.1 | |||
XM_011512039.2 | 294 | Intron | XP_011510341.1 | |||
XM_011512040.2 | 294 | Intron | XP_011510342.1 | |||
XM_017005138.1 | 294 | UTR 5 | XP_016860627.1 | |||
XM_017005139.1 | 294 | Intron | XP_016860628.1 | |||
XM_017005140.1 | 294 | Intron | XP_016860629.1 |
SAG - S-antigen visual arrestin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |